2-Fluoro-6-formylbenzeneboronic acid ≥98%

Supplier: Ambeed
Warning

A569305-5G A569305-10G
77747-173EA 136 USD
77747-173 77747-172
2-Fluoro-6-formylbenzeneboronic acid ≥98%
2-Fluoro-6-formylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MDL Number: MFCD19981524
CAS Number: 1938062-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR