5-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one ≥95%

Supplier: Ambeed
Warning

A580256-250MG A580256-100MG A580256-1G
77598-582EA 108.65 USD
77598-582 77617-448 77749-196
5-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one ≥95%
5-Bromo-2-methyl-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MDL Number: MFCD20482681
CAS Number: 1100509-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR