Amsacrine hydrochloride 98+%

Supplier: Ambeed
Danger

Synonyms: m-AMSA, N-(4-(Acridin-9-ylamino)-3-methoxyphenyl)methanesulphonamide hydrochloride, 4-(9-Acridinylamino)-N-(methanesulphonyl)-m-anisidine hydrochloride, Amsacrine HCl

A580248-250MG A580248-5G
77627-194EA 51.5 USD
77627-194 77878-381
Amsacrine hydrochloride 98+%
Amsacrine hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₃O₃S·HCl
MW: 429.92 g/mol
MDL Number: MFCD07799963
CAS Number: 54301-15-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR