4-Bromo-1-methyl-1H-pyrrole-2-carbonitrile ≥95%

Supplier: Ambeed
Warning

A568189-100MG A568189-250MG
77891-139EA 61 USD
77891-139 77891-140
4-Bromo-1-methyl-1H-pyrrole-2-carbonitrile ≥95%
4-Bromo-1-methyl-1H-pyrrole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₅BrN₂
MDL Number: MFCD18917074
CAS Number: 1289207-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR