(S)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile ≥98%

Supplier: Ambeed
Warning

Synonyms: Ruxolitinib (S enantiomer)

A579847-10MG A579847-1MG A579847-5MG
77617-438EA 90.64 USD
77617-438 77617-440 77617-442
(S)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile ≥98%
(S)-3-(4-(7H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₆
MW: 306.37 g/mol
MDL Number: MFCD13184797
CAS Number: 941685-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR