6-Chloro-2-methyl-4-pyrimidinol ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-4-hydroxy-2-methylpyrimidine, 6-Chloro-2-methylpyrimidin-4(1H)-one

A568117-5G A568117-1G
77572-806EA 104.03 USD
77572-806 77749-169
6-Chloro-2-methyl-4-pyrimidinol ≥98%
6-Chloro-2-methyl-4-pyrimidinol

Precaución: for Research Use Only

Formula: C₅H₅ClN₂O
MW: 144.558 g/mol
MDL Number: MFCD00030773
CAS Number: 17551-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR