2-Bromo-6-fluoro-4-methylbenzaldehyde ≥98%

Supplier: Ambeed
Warning

A568089-1G A568089-5G
77891-137EA 24 USD
77891-137 77891-138
2-Bromo-6-fluoro-4-methylbenzaldehyde ≥98%
2-Bromo-6-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD22056736
CAS Number: 1370025-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR