5-tert-Butyl-m-xylene ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-tert-Butyl-3,5-dimethylbenzene, 1-(tert-Butyl)-3,5-dimethylbenzene

A568072-100G A568072-25G A568072-500G A568072-1000G
77572-804EA 19.57 USD
77572-804 77602-392 77602-394 77627-174
5-tert-Butyl-m-xylene ≥98%
5-tert-Butyl-m-xylene

Precaución: for Research Use Only

Formula: C₁₂H₁₈
MW: 162.27 g/mol
MDL Number: MFCD00008832
CAS Number: 98-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR