5-Bromopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester ≥98%

Supplier: Ambeed
Warning

A579690-1G A579690-250MG A579690-5G
77891-151EA 52 USD
77891-151 77891-152 77891-153
5-Bromopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester ≥98%
5-Bromopyrrolo[2,3-b]pyridine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O₂
MDL Number: MFCD08741534
CAS Number: 928653-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR