cis-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A567836-100MG A567836-250MG
77686-447EA 213 USD
77686-447 77686-448
cis-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-3-carboxylic acid ≥98%
cis-1-(tert-Butoxycarbonyl)-4-fluoropyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₄
MW: 233.23 g/mol
MDL Number: MFCD23703411
CAS Number: 1427501-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR