Fmoc-2,3-Dimethylphe ≥95%

Supplier: Ambeed
Warning

A579194-1G A579194-250MG
77783-473EA 264 USD
77783-473 77783-474
Fmoc-2,3-Dimethylphe ≥95%
Fmoc-2,3-Dimethylphe

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₄
MDL Number: MFCD09842054
CAS Number: 1270295-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR