1-(4-Chlorophenyl)guanidine hydrochloride ≥96%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenylguanidine hydrochloride

A578940-250MG A578940-1G
77749-191EA 63 USD
77749-191 77749-190
1-(4-Chlorophenyl)guanidine hydrochloride ≥96%
1-(4-Chlorophenyl)guanidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂N₃
MW: 206.07 g/mol
MDL Number: MFCD00209839
CAS Number: 14279-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR