2-(3-((tert-Butoxycarbonyl)amino)cyclobutyl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-[3-(tert-butoxycarbonylamino)cyclobutyl]acetic acid

A566931-100MG A566931-250MG A566931-1G
77686-444EA 50 USD
77686-444 77686-445 77901-014
2-(3-((tert-Butoxycarbonyl)amino)cyclobutyl)acetic acid ≥98%
2-(3-((tert-Butoxycarbonyl)amino)cyclobutyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
MDL Number: MFCD28501915
CAS Number: 1638772-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR