(1S,3'R,4'S,5'S,6'R)-6-(4-Ethylbenzyl)-6'-(hydroxymethyl)-3',4',5',6'-tetrahydro-3H-spiro[isobenzofuran-1,2'-pyran]-3',4',5'-triol hydrate ≥99%

Supplier: Ambeed
Warning

Synonyms: Tofogliflozin (hydrate)

A566836-10MG A566836-5MG
77747-167EA 132 USD
77747-167 77747-168
(1S,3'R,4'S,5'S,6'R)-6-(4-Ethylbenzyl)-6'-(hydroxymethyl)-3',4',5',6'-tetrahydro-3H-spiro[isobenzofuran-1,2'-pyran]-3',4',5'-triol hydrate ≥99%
(1S,3'R,4'S,5'S,6'R)-6-(4-Ethylbenzyl)-6'-(hydroxymethyl)-3',4',5',6'-tetrahydro-3H-spiro[isobenzofuran-1,2'-pyran]-3',4',5'-triol hydrate

Precaución: for Research Use Only

Formula: C₂₂H₂₈O₇
MW: 404.45 g/mol
MDL Number: MFCD28385880
CAS Number: 1201913-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR