2'-Chloro-5'-hydroxyacetophenone ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Chloro-5-hydroxyphenyl)ethanone
A578733-1G
A578733-100MG
77747-200EA
168
USD
77747-200
77747-199
2'-Chloro-5'-hydroxyacetophenone ≥97%
2'-Chloro-5'-hydroxyacetophenone
Precaución: for Research Use Only
Formula:
C₈H₇ClO₂ MW: 170.593 g/mol |
MDL Number:
MFCD18393141 CAS Number: 58020-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...