1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile ≥95%

Supplier: Ambeed
Warning

A566589-1G A566589-250MG
77673-514EA 80 USD
77673-514 77673-516
1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile ≥95%
1H-Pyrrolo[3,2-c]pyridine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MDL Number: MFCD09965899
CAS Number: 1040682-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR