Dibutylamine hydrochloride ≥98%

Supplier: Ambeed
Warning

A566224-100G A566224-500G
77880-221EA 65 USD
77880-221 77880-222
Dibutylamine hydrochloride ≥98%
Dibutylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₂₀ClN
MW: 165.71 g/mol
MDL Number: MFCD00034885
CAS Number: 6287-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR