(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-methoxyphenyl)propanoic acid ≥97%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(2-OMe)-OH

A578465-250MG A578465-1G
77747-197EA 40 USD
77747-197 77747-196
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-methoxyphenyl)propanoic acid ≥97%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(2-methoxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.33 g/mol
MDL Number: MFCD01860640
CAS Number: 143415-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR