5-(Chloromethyl)-2-fluoropyridine ≥98%

Supplier: Ambeed
Danger

A566042-1G A566042-250MG A566042-5G
77893-146EA 72 USD
77893-146 77893-147 77893-148
5-(Chloromethyl)-2-fluoropyridine ≥98%
5-(Chloromethyl)-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MDL Number: MFCD10697563
CAS Number: 315180-15-5
UN: 1760
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR