(4S)-Methyl-[1,3,2]dioxathiolane 2,2-dioxide ≥95%

Supplier: Ambeed
Danger

A578241-1G A578241-250MG A578241-5G
77622-374EA 122.57 USD
77622-374 77783-469 77783-470
(4S)-Methyl-[1,3,2]dioxathiolane 2,2-dioxide ≥95%
(4S)-Methyl-[1,3,2]dioxathiolane 2,2-dioxide

Precaución: for Research Use Only

Formula: C₃H₆O₄S
MDL Number: MFCD26131320
CAS Number: 174953-30-1
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR