5-Methoxy-2-methylpyridine ≥98%

Supplier: Ambeed
Warning

A565673-1G A565673-250MG
77878-363EA 81 USD
77878-363 77878-364
5-Methoxy-2-methylpyridine ≥98%
5-Methoxy-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₉NO
MDL Number: MFCD09833595
CAS Number: 55270-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR