4-Methoxy-3-pyridineboronic acid hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A565116-1G
A565116-250MG
A565116-5G
77884-384EA
34
USD
77884-384
77884-385
77884-386
4-Methoxy-3-pyridineboronic acid hydrochloride ≥98%
4-Methoxy-3-pyridineboronic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₉BClNO₃ |
MDL Number:
MFCD04113800 CAS Number: 874959-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...