Nucleozin ≥99%

Supplier: Ambeed
Warning

A577855-10MG A577855-5MG
77747-194EA 87 USD
77747-194 77747-195
Nucleozin ≥99%
Nucleozin

Precaución: for Research Use Only

Formula: C₂₁H₁₉ClN₄O₄
MDL Number: MFCD01426660
CAS Number: 341001-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR