4-Ethyl-2-nitroaniline ≥97%

Supplier: Ambeed
Danger

A577533-1G A577533-250MG
77572-844EA 71 USD
77572-844 77747-193
4-Ethyl-2-nitroaniline ≥97%
4-Ethyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00034069
CAS Number: 3663-35-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR