4-Chloro-5-methoxy-2-methylpyrimidine ≥95%

Supplier: Ambeed
Warning

A577478-1G A577478-250MG
77594-752EA 560.68 USD
77594-752 77613-788
4-Chloro-5-methoxy-2-methylpyrimidine ≥95%
4-Chloro-5-methoxy-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MDL Number: MFCD00223701
CAS Number: 698-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR