1-(((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-6-methoxyquinolin-7-yl)oxy)methyl)cyclopropanamine dihydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Catequentinib HCl

A564645-1MG A564645-5MG
77598-576EA 28.7 USD
77598-576 77598-578
1-(((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-6-methoxyquinolin-7-yl)oxy)methyl)cyclopropanamine dihydrochloride ≥98%
1-(((4-((4-Fluoro-2-methyl-1H-indol-5-yl)oxy)-6-methoxyquinolin-7-yl)oxy)methyl)cyclopropanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₄Cl₂FN₃O₃
MW: 480.36 g/mol
MDL Number: MFCD31617980
CAS Number: 1360460-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR