(1,5-Dimethyl-1H-pyrazol-4-yl)methanol ≥95%

Supplier: Ambeed
Warning

A564578-1G A564578-250MG
77673-510EA 216 USD
77673-510 77882-359
(1,5-Dimethyl-1H-pyrazol-4-yl)methanol ≥95%
(1,5-Dimethyl-1H-pyrazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MDL Number: MFCD02253731
CAS Number: 103946-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR