4-Bromo-1H-indazol-6-amine ≥95%

Supplier: Ambeed
Warning

A564413-250MG A564413-100MG A564413-1G
77594-722EA 47.15 USD
77594-722 77684-417 77684-418
4-Bromo-1H-indazol-6-amine ≥95%
4-Bromo-1H-indazol-6-amine

Precaución: For research use only.

Formula: C₇H₆BrN₃
MW: 212.04 g/mol
MDL Number: MFCD07781322
CAS Number: 885518-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR