1-Methyl-3-trifluoromethylpyrazole-4-boronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

A564323-250MG A564323-1G
77594-720EA 14.35 USD
77594-720 77673-508
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid ≥95%
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BF₃N₂O₂
MW: 193.92 g/mol
MDL Number: MFCD13619875
CAS Number: 1138450-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR