1-(3-Bromophenyl)-1-methylethylamine ≥98%

Supplier: Ambeed
Warning

A576063-1G A576063-250MG
77893-162EA 103 USD
77893-162 77893-163
1-(3-Bromophenyl)-1-methylethylamine ≥98%
1-(3-Bromophenyl)-1-methylethylamine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN
MDL Number: MFCD11109722
CAS Number: 74702-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR