6-Bromo-7-methyl-1H-indole ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Danger
|
A564131-250MG
A564131-100MG
77684-416EA
27
USD
77684-416
77886-379
6-Bromo-7-methyl-1H-indole ≥98%, Ambeed.Inc
6-Bromo-7-methyl-1H-indole
Precaución: For research use only.
Formula:
C₉H₈BrN |
MDL Number:
MFCD09027015 CAS Number: 1000343-89-4 UN: 2923 ADR: 8, 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...