(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol ≥98%

Supplier: Ambeed
Danger

Synonyms: Bekanamycin

A575687-1MG A575687-100MG
77669-514EA 21 USD
77669-514 77880-239
(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol ≥98%
(2R,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4-diol

Precaución: for Research Use Only

Formula: C₁₈H₃₇N₅O₁₀
MDL Number: MFCD01683541
CAS Number: 4696-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR