4-Nitro-piperidine-1-carboxylic acid tert-butyl ester ≥98%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-nitropiperidine-1-carboxylate

A563858-1G A563858-250MG
77594-716EA 55.35 USD
77594-716 77684-415
4-Nitro-piperidine-1-carboxylic acid tert-butyl ester ≥98%
4-Nitro-piperidine-1-carboxylic acid tert-butyl ester

Precaución: For research use only.

Formula: C₁₀H₁₈N₂O₄
MW: 230.261 g/mol
MDL Number: MFCD16618559
CAS Number: 1228630-89-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR