4-Cyano-3-(trifluoromethyl)phenylboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: (4-Cyano-3-(trifluoromethyl)phenyl)boronic acid

A575374-100MG A575374-1G
77882-375EA 14 USD
77882-375 77882-376
4-Cyano-3-(trifluoromethyl)phenylboronic acid ≥95%
4-Cyano-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₃NO₂
MW: 214.937 g/mol
MDL Number: MFCD10697412
CAS Number: 915299-32-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR