Thieno[2,3-b]pyridin-4-ol ≥96%

Supplier: Ambeed
Warning

A563700-250MG A563700-1G
77891-124EA 64 USD
77891-124 77891-123
Thieno[2,3-b]pyridin-4-ol ≥96%
Thieno[2,3-b]pyridin-4-ol

Precaución: for Research Use Only

Formula: C₇H₅NOS
MDL Number: MFCD18802558
CAS Number: 65075-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR