2-(Bromomethyl)-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(2-(Bromomethyl)-4-(trifluoromethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A574979-1G A574979-100MG A574979-250MG
77686-465EA 317 USD
77686-465 77900-939 77900-739
2-(Bromomethyl)-4-(trifluoromethoxy)phenylboronic acid pinacol ester ≥98%
2-(Bromomethyl)-4-(trifluoromethoxy)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₇BBrF₃O₃
MDL Number: MFCD09475917
CAS Number: 957066-13-6
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR