5-Bromo-4-chloro-3-indolyl acetate ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4-chloroindoxyl acetate, 5-Bromo-4-chloro-1H-indol-3-yl acetate, BCDA, 5-Bromo-4-chloro-3-indolyl acetate, 3-Acetoxy-4-chloro-5-bromoindole

A574954-1G A574954-100MG A574954-250MG
77598-580EA 316.73 USD
77598-580 77627-188 77627-190
5-Bromo-4-chloro-3-indolyl acetate ≥98%
5-Bromo-4-chloro-3-indolyl acetate

Precaución: for Research Use Only

Formula: C₁₀H₇BrClNO₂
MW: 288.525 g/mol
MDL Number: MFCD00037932
CAS Number: 3252-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR