3-exo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol ≥98%

Supplier: Ambeed
Warning

Synonyms: Pseudotropine

A574606-250MG A574606-10G
77747-190EA 5 USD
77747-190 77747-189
3-exo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol ≥98%
3-exo-8-Methyl-8-azabicyclo[3.2.1]octan-3-ol

Precaución: for Research Use Only

Formula: C₈H₁₅NO
MW: 141.21 g/mol
MDL Number: MFCD00078060
CAS Number: 135-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR