(R)-3-Hydroxy-2-methylpropanoic acid ≥98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: (R)-2-Hydroxymethylpropanoic acid

A574289-1G A574289-250MG
77617-424EA 524.27 USD
77617-424 77617-426
(R)-3-Hydroxy-2-methylpropanoic acid ≥98% (by HPLC)
(R)-3-Hydroxy-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.10 g/mol
MDL Number: MFCD06656409
CAS Number: 1910-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR