(6-Fluoro-1H-benzimidazol-2-yl)methanol ≥98%

Supplier: Ambeed
Warning

A573610-250MG A573610-1G
77886-394EA 10 USD
77886-394 77886-393
(6-Fluoro-1H-benzimidazol-2-yl)methanol ≥98%
(6-Fluoro-1H-benzimidazol-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₇FN₂O
MDL Number: MFCD09261475
CAS Number: 39811-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR