(3,5-Difluoro-4-(trifluoromethyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A573473-100MG A573473-250MG
77686-458EA 48 USD
77686-458 77686-459
(3,5-Difluoro-4-(trifluoromethyl)phenyl)boronic acid ≥95%
(3,5-Difluoro-4-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₄BF₅O₂
MW: 225.90 g/mol
MDL Number: MFCD28291812
CAS Number: 1459778-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR