9,9'-Bis([1,1'-biphenyl]-3-yl)-3,3'-bi-9H-carbazole ≥98%
Supplier: Ambeed
|
Warning
|
A562637-1G
A562637-100MG
A562637-250MG
77622-368EA
200.85
USD
77622-368
77783-460
77783-461
9,9'-Bis([1,1'-biphenyl]-3-yl)-3,3'-bi-9H-carbazole ≥98%
9,9'-Bis([1,1'-biphenyl]-3-yl)-3,3'-bi-9H-carbazole
Precaución: for Research Use Only
Formula:
C₄₈H₃₂N₂ |
MDL Number:
MFCD32664645 CAS Number: 1352040-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...