Benzyl (S)-(2-((2-((5-guanidino-1-((4-methyl-2-oxo-2H-chromen-7-yl)amino)-1-oxopentan-2-yl)amino)-2-oxoethyl)amino)-2-oxoethyl)carbamate acetate ≥98%

Supplier: Ambeed
Warning

Synonyms: Z-Gly-Gly-Arg-AMC acetate

A573033-10MG A573033-5MG
77749-179EA 80 USD
77749-179 77749-180
Benzyl (S)-(2-((2-((5-guanidino-1-((4-methyl-2-oxo-2H-chromen-7-yl)amino)-1-oxopentan-2-yl)amino)-2-oxoethyl)amino)-2-oxoethyl)carbamate acetate ≥98%
Benzyl (S)-(2-((2-((5-guanidino-1-((4-methyl-2-oxo-2H-chromen-7-yl)amino)-1-oxopentan-2-yl)amino)-2-oxoethyl)amino)-2-oxoethyl)carbamate acetate

Precaución: for Research Use Only

Formula: C₃₀H₃₇N₇O₉
MW: 639.66 g/mol
MDL Number: MFCD29472228
CAS Number: 2070009-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR