3-(2,5-Dimethylphenyl)propionic acid ≥95%

Supplier: Ambeed
Warning

A562244-100MG A562244-250MG
77613-756EA 15.45 USD
77613-756 77901-991
3-(2,5-Dimethylphenyl)propionic acid ≥95%
3-(2,5-Dimethylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06823972
CAS Number: 25173-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR