1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole 98+%

Supplier: Ambeed
Warning

A572090-100MG A572090-1G A572090-250MG A572090-5G
77893-151EA 60 USD
77893-151 77893-152 77893-153 77893-154
1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole 98+%
1-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-benzo[d]imidazole

Precaución: for Research Use Only

Formula: C₁₄H₁₉BN₂O₂
MDL Number: MFCD16995903
CAS Number: 1107627-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR