4-(2-Carboxyethyl)-4-nitroheptanedioic acid ≥98%

Supplier: Ambeed
Warning

A571959-100G A571959-5G
77572-820EA 291 USD
77572-820 77749-176
4-(2-Carboxyethyl)-4-nitroheptanedioic acid ≥98%
4-(2-Carboxyethyl)-4-nitroheptanedioic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₈
MW: 277.23 g/mol
MDL Number: MFCD00077684
CAS Number: 59085-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR