trans-Cyclobutane-1,3-diol ≥98%

Supplier: Ambeed
Warning

A571795-100MG A571795-1G
77783-467EA 223 USD
77783-467 77783-468
trans-Cyclobutane-1,3-diol ≥98%
trans-Cyclobutane-1,3-diol
Formula: C₄H₈O₂
MDL Number: MFCD28139477
CAS Number: 1332482-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR