(R)-1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarbaldehyde acetic acid salt ≥98%

Supplier: Ambeed
Warning

Synonyms: (R)-(-)-Gossypol acetic acid

A571691-10MG A571691-5MG
77747-180EA 89 USD
77747-180 77747-181
(R)-1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarbaldehyde acetic acid salt ≥98%
(R)-1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarbaldehyde acetic acid salt

Precaución: for Research Use Only

Formula: C₃₂H₃₄O₁₀
MW: 578.61 g/mol
MDL Number: MFCD00058385
CAS Number: 866541-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR