N-(4-Pentynyl)phthalimide ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(Pent-4-yn-1-yl)isoindoline-1,3-dione

A571329-1G A571329-5G
77673-532EA 20 USD
77673-532 77884-396
N-(4-Pentynyl)phthalimide ≥98%
N-(4-Pentynyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD06798110
CAS Number: 6097-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR