3,5-Dimethylpyridine-4-boronic acid pinacol ester 98+%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

A561411-1G A561411-100MG
77572-774EA 550 USD
77572-774 77886-375
3,5-Dimethylpyridine-4-boronic acid pinacol ester 98+%
3,5-Dimethylpyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₀BNO₂
MDL Number: MFCD10566506
CAS Number: 1032358-02-3
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR